Order Entry
Poland
ContactUsLinkComponent
Thermosafe Cool Boxes
Thermosafe Cool Boxes
Catalog # 216-7001
Supplier:  HONIGBERG
undefined
Thermosafe Cool Boxes
Catalog # 216-7001
Supplier:  HONIGBERG

Specifications

  • Capacity:
    75 l
  • Description:
    Cooling box Thermosafe
  • Dimensions:
    585×560×510 mm (ext); 455×440×410 mm (int)
  • Pk:
    1

Specifications

About this item

PE, grey.

  • Insulated with 5,75 cm thick plastic foam
  • Hinged lid with robust hinges and metal catch, one sturdy carrying handle with the 35 litre model and two with the 75 litre model
  • Aluminium roller base, with 4 moving rollers, available separately

Large capacity cool boxes for the long-term temperature control of temperature-sensitive materials and samples. Cooled with cooling pouches 216-7010 to 216-7013 or dry ice.