Bottles, wide neck, with screw cap, Azlon®
PP, high clarity, with PP screw cap.
- Leakproof, liner-less design
- Clear crisp printed graduations show millilitres and fluid ounces
- Printed with large white 'write on' panel for easy labelling
- Autoclavable at 121 °C
- Smooth inner contour for efficient pouring and ease of cleaning
Ideal for both liquids and solids, perfect for microbiological sampling.
When autoclaving bottles always ensure the caps are completely loosened or removed to prevent accidental implosion.
Frequently Bought Together