PROTEIN KW-800 and KW400 series HPLC columns are specifically designed for protein analysis by aqueous size exclusion chromatography (GFC). PROTEIN LW-803 and LW-403 4D are designed for antibody analysis. They are packed with porous silica particles coated with a hydrophilic copolymer. They fulfill USP L20, L33, and L59 requirements.
Standard columns PROTEIN KW-800 series
- Silica-based packing material for aqueous SEC analysis.
- Suitable for the analysis of proteins and enzymes.
- There are three pore sizes available for different molecular weight ranges.
High performance semi-micro columns PROTEIN KW400 series
- Reduced particle size for enhanced column performance.
- Faster separation and higher sensitivity than PROTEIN KW-800 columns.
- There are four pore sizes available for different molecular weight ranges.
PROTEIN LW columns for antibody drug analysis
- Pore size specifically controlled for analyzing proteins with a molecular weight of several hundred of thousand.
- High performance analysis of monoclonal antibodies (mAb) in therapeutic drugs.
- High lot-to-lot reproducibility.
- Standard and semi-micro column dimension available.
* Estimated value.
- Catalog No:
- SHOWF6989403
- 554-1739
- 554-1737
- 554-1743
- 554-5779
- 554-1740
- 554-5778
- SHOWF6700134
- 554-1741
- 554-1744
- 554-1735
- 554-1736
- 554-1742
- Description:
- PROTEIN LW-403 4D
- PROTEIN KW-G 6B
- PROTEIN KW-804
- PROTEIN KW405-4F
- PROTEIN LW-G 6B
- PROTEIN KW402.5-4F
- PROTEIN LW-803
- PROTEIN LS-G 4J
- PROTEIN KW403-4F
- PROTEIN KW400G-4A
- PROTEIN KW-802.5
- PROTEIN KW-803
- PROTEIN KW404-4F
- Max. pressure:
- 5,0 MPa (50 bar)
- 7,0 MPa (70 bar)
- 10 MPa (100 bar)
- 10 MPa (100 bar)
- 5,0 MPa (50 bar)
- 5,0 MPa (50 bar)
- 7,0 MPa (70 bar)
- pH range:
- 3 - 7,5
- 3 - 7,5
- 3 - 7,5
- 3 - 7,5
- 3 - 7,5
- 3 - 7,5
- 3 - 7,5
- 3 - 7,5
- 3 - 7,5
- Temperature limit:
- 4 - 45 °C
- 4 - 45 °C
- 5 - 45 °C
- 5 - 45 °C
- 5 - 45 °C
- 5 - 45 °C
- 4 - 45 °C
- 4 - 45 °C
- 5 - 45 °C
- USP code:
- L20, L33, L59
- L20, L33, L59
- L20, L33, L59
- L20, L33, L59
- L20, L33, L59
- L20, L33, L59
- L20, L33, L59
- L20, L33, L59
- L20, L33, L59