Order Entry
Poland
ContactUsLinkComponent
3-Amino-1H-1,2,4-triazole, Sigma-Aldrich®
3-Amino-1H-1,2,4-triazole, Sigma-Aldrich®
Supplier:  Merck
Certificates
3-Amino-1H-1,2,4-triazole, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    280 g/l (25 °C)
  • Melting Point:
    149 - 152 °C
  • Molar Mass:
    84.08 g/mol
  • Bulk Density:
    650 kg/m3
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Density:
    1.138 g/cm3 (20 °C)
  • pH:
    6.4 - 7.4 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    225 °C

Specification Test Results

Related Information

Frequently Bought Together