Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α,α-Dichloropropiophenone
α,α-Dichloropropiophenone
α,α-Dichloropropiophenone
8.14489.0005
:
Merck
:
α,α-Dichloropropiophenone
8.14489.0005
:
Merck
:
Pack type:
Glass bottle
Pk:
5 ml
Seq:
2210
:
:
110…114 °C (19 hPa)
:
1.26 g/cm³ (20 °C)
:
203.07 g/mol
:
57169-51-4
:
C₆H₅C(O)CCl₂CH₃