Order Entry
Poland
Orders LinkContactUsLinkComponent
MultiScreen® PCR<sub>µ96</sub> Filter Plates
MultiScreen® PCRµ96 Filter Plates
New Product
Supplier:   MILLIPORE
MultiScreen® PCR<sub>µ96</sub> Filter Plates
Supplier:  MILLIPORE
New Product
Certificates
MultiScreen® PCR<sub>µ96</sub> Filter Plates
MultiScreen® PCRµ96 Filter Plates
2 Options Available Below
Product Details & Documents
MultiScreen® PCRµ96 filter plate is a small volume, 96-well device for the purification of polymerase chain reaction (PCR) products. The PCRμ96 plate enables purification of both small and large volume PCR reactions (20 to 100 μl) with the recovery of purified products in as little as 20 μl of solution.
  • Offers fast, automatable solutions for high-throughput PCR purification
  • Provides high purity and high recovery
The vacuum-based, size exclusion separation effectively removes contaminating salts, unincorporated dNTPs, and primers from PCR reactions in less than 15 minutes. The purified DNA is suitable for downstream applications such as sequencing, genotyping, or micro arraying. Additionally, the automation-friendly design of the MultiScreen® PCRµ96 plate makes it ideal for high throughput applications.
Application: MultiScreen® PCRµ96 filter plate has been used in natural product screening, total drug analysis, protein precipitation, bead cleavage, solid-phase synthesis, and solubility assay. It has also been used in the purification of polymerase chain reaction (PCR) products.
Well maximum volume: 175 µl
Product Family Options
Product Information
  • Material
    Well volume
    Recommended working volume
    No. of wells
    Sterile
    Availability
    Price
  • MILYLSKMPCR10
    Filter plate, no lid, low binding surface
    Filter plate, no lid, low binding surface
    Polystyrene
    50 - 250 µl
    100 - 150 µl
    96
    -
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 4 342,50 zł
    4 342,50 zł
    1 * 10 SZT
       4 342,50 zł
    Specifications:
    New Product
    New Product
  • MILYLSKMPCR50
    Filter plate, no lid, low binding surface
    Filter plate, no lid, low binding surface
    Polystyrene
    50 - 250 µl
    100 - 150 µl
    96
    -
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 14 467,50 zł
    14 467,50 zł
    1 * 1 SZT
       14 467,50 zł
    Specifications:
    New Product
    New Product