Amicon® Stirred Cell Selector Valve
Catalog # MILX6003-M
Supplier: Merck
New Product
Specifications
- Description:Amicon® Stirred cell selector valve
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Valve with sliding control for instant switching from concentration to diafiltration, or switching gas and liquid lines simultaneously. Simplifies operation and avoids the need for multiple T-fittings and valves.
- Device Material: Acetal, Buna-N and nylon
- Chemically sanitizable
- Dimension: 75×20×80 mm
The selector valve provides a quick and simple way to switch the Amicon Stirred Cell between concentration and diafiltration modes without interrupting system operation.
Application: The selector valve provides a quick and simple way to switch the Amicon Stirred Cell between concentration and diafiltration modes without interrupting system operation.
Key Applications: Concentration and diafiltration.