Multi Channel Pipettes, Variable Volume, Labmate Pro
8-channel pipettor, 20 - 200 µl
HTLA6283
: HTL
Labmate variable volume pipettors have been the highlight of the HTL product range. Their outstandingly accurate and precise pipetting mechanism, enclosed in ergonomic, well balanced and robust handle made Labmate the pipettor of choice for many laboratory professionals worldwide.
- Contoured shape of the handle, equipped with a finger-hook guarantee correct grip reducing wrist strain and fatigue
- Low pipettor weight and soft springs system provides comfort at work, even during long pipetting series
- Reliable sealing system ensures reliability of results
- Built-in user-recalibration system allows matching of the pipettor for your specific application
- Fully autoclavable
- Ejector with infinitely variable length adjustment makes it easier for different tips
- Rotating multi-channel module allows free rotation of 360°
The most recent version of this popular pipettor - Labmate Pro combines so appreciated traditional design with the new pipetting mechanism and more durable components, making the daily pipetting routine an effortless and reliable experience.
- Volume:20 - 200 µl
- Description:8-channel pipettor