Organic reference standard, Acrylonitrile 5000 µg/ml in methanol, Supelco®
Catalog # SUPL40003
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- CAS No.:107-13-1
- Description:Acrylonitrile 5,000 µg/ml in methanol
- Formula:C₃H₃N
- Description:Acrylonitrile