3,3'-Dihydroxybenzidine 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
- CAS Number:2373-98-0
- Formula:C₁₂H₁₂N₂O₂
- MDL Number:MFCD00039149
- Molecular Weight:216.24 g/mol
- Storage Temperature:Temperatura otoczenia
- EINECS:220-190-0
3 Options Available Below
Product Details & Documents
Synonyms: 4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
- CAS Number:2373-98-0
- Formula:C₁₂H₁₂N₂O₂
- MDL Number:MFCD00039149
- Molecular Weight:216.24 g/mol
- Storage Temperature:Temperatura otoczenia
- EINECS:220-190-0
Specifications for Product Family
Specification Test Results
- Appearance:white to brown powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



