Polyvinylpyrrolidone K30, meets analytical specification of Ph.Eur.
New Product
:9003-39-8
:
:(C₆H₉NO)n
:MFCD01076626
:13,07783
:Ambient
- Appearance:white or yellowish-white powder or flakes
- Infrared spectrum:Conforms
- Peroxides:≤400 ppm (type A as H2O2)
- Nitrogen compounds (as N):11.5 to 12.8 %
- Viscosity:28 to 32 (expressed as K-value)
Frequently Bought Together