Order Entry
Poland
ContactUsLinkComponent
 
Bismuth(III) nitrate 0,01 mol/L
Catalog # ACSDCH8006005.1L
Supplier:  CPACHEM
New Product
CAS Number:  
Bismuth(III) nitrate 0,01 mol/L
Catalog # ACSDCH8006005.1L
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :395
Boiling Pt:100 °C
Density:1 g/cm³
CAS Number:10361-44-1
Formula:Bi(NO₃)₃·xH₂O
MDL Number:MFCD00003460