Order Entry
Poland
ContactUsLinkComponent
 
Flammable
Corrosive
Iodine monobromide solution 20 g/l in acetic acid (99%) Reag. Ph. Eur. 1045901
Catalog # 87714.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Corrosive
Iodine monobromide solution 20 g/l in acetic acid (99%) Reag. Ph. Eur. 1045901
Catalog # 87714.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :206.81 g/mol
Storage Temperature:Ambient
EINECS:232-159-9
UN:2789
ADR:8 (3),II
 :GHS02
 :GHS05
CAS Number:7789-33-5
Formula:IBr
MDL Number:MFCD00011353