Order Entry
Poland
ContactUsLinkComponent
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) 40 mg, capsules alkaline phosphatase substrate, Sigma-Aldrich®
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) 40 mg, capsules alkaline phosphatase substrate, Sigma-Aldrich®
Catalog # SIALP5744-25CAP
Supplier:  Merck
CAS Number:  
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) 40 mg, capsules alkaline phosphatase substrate, Sigma-Aldrich®
Catalog # SIALP5744-25CAP
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25
  • Seq:
    0025

Specifications

About this item

 :
Melting Pt:300 °C
 :M-Clarity™ quality level = MQ200
 :371.144 g/mol
Storage Temperature:Freezer
EINECS:224-246-5
CAS Number:333338-18-4
 :4264-83-9
Formula:C₆H₄NNa₂O₆P·6H₂O
MDL Number:MFCD00066288