Supelco® TraceCERT® Organic Reference Standard, Ethirimol
Catalog # 93638-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- CAS No.:23947-60-6
- Description:Ethirimol
- Storage conditions:15...25 °C
- Hazard class description:H312
- MDL No.:MFCD00055519
- M-Clarity quality level:MQ300
- Molecular weight:209,29
Specifications
About this item
Ethirimol is the ethylamino isomer of dimethirimol that belongs to a group of pyrimidine fungicides. This systemic fungicide acts by inhibiting nucleic acid synthesis and is used as a seed treatment to control powdery mildew in spring barley and wheat. Ethirimol is not approved for use in European Union (EU) as per Regulation (EC) No 1107/2009. But a default maximum residue level (MRLs) from 0,01 to 2 mg/kg is applied for products of plant and animal origin according to Regulation (EU) 2021/1531. Ethirimol has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP) run within the EU and EFTA in/on products of plant origin.
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P280 - P302 + P352 + P312 - P362 + P364 - P501