Order Entry
Poland
ContactUsLinkComponent
Supelco® Organic Reference Standard, 3-Carboxy-4-methyl-5-propyl-2-furanpropanoic acid
Supelco® Organic Reference Standard, 3-Carboxy-4-methyl-5-propyl-2-furanpropanoic acid
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, 3-Carboxy-4-methyl-5-propyl-2-furanpropanoic acid
Supplier:  Merck

Specifications

  • CAS No.:
    86879-39-2
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD08436079
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-Carboxy-4-methyl-5-propyl-2-furanpropionic acid; CMPF; Propylurofuranic acid
  • Molecular weight:
    240,25

Specifications

Frequently Bought Together