Z-DEVD-aminoluciferin
1 mg
Catalog # AATB13211
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Z-DEVD-aminoluciferin is a luminogenic substrate that measures caspase-3 and -7 activities in apoptotic cells.
- Stable for 24 months
Quality tested in-house
: For Research Use Only
- Molecular Weight:871.89 g/mol
Specifications
Specifications
- Pk:1 mg