Anti-OR2W3 Rabbit Polyclonal Antibody
Catalog # ANTIA100914-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Olfactory receptor 2W3
- Antigen symbol:OR2W3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q7Z3T1
- Antigen synonyms:Olfactory receptor family 2 subfamily W member 8 pseudogene|Olfactory receptor family 2 subfamily W member 3 pseudogene|OR2W3|OR2W3P|OR2W3_HUMAN|OR2W8P|Olfactory receptor 2W8|Olfactory receptor OR1-49|Olfactory receptor family 2 subfamily W member 3
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:34 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human OR2W3 (amino acids 141-190)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to OR2W3 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: OR2W3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human