Order Entry
Poland
ContactUsLinkComponent
 
Anti-CD38 Mouse Monoclonal Antibody [Clone: HIT2] (PE (Phycoerythrin))
  ANTIA86185-100
New Product
undefined
Anti-CD38 Mouse Monoclonal Antibody [Clone: HIT2] (PE (Phycoerythrin))
  ANTIA86185-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Acute lymphoblastic leukemia cells antigen CD38
  • Antigen symbol:
    CD38
  • Clonality:
    Monoclonal
  • Clone:
    HIT2
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28907
  • Antigen synonyms:
    ADPRC 1|ADP-ribosyl cyclase 1|CD38 antigen|ADPRC1|cADPr hydrolase 1|ADP ribosyl cyclase/cyclic ADP-ribose hydrolase|ADP ribosyl cyclase|ADP ribosyl cyclase 1|CD 38
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human thymocytes in foetus
  • Purification:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

 

 

Mouse monoclonal [HIT2] antibody to CD38 (PE) for Flow Cytometry with samples derived from Human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human