Anti-Integrin alpha 4 Mouse Monoclonal Antibody [Clone: 9F10]
Catalog # ANTIA86038-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:269C wild type
- Antigen symbol:Integrin alpha 4
- Clonality:Monoclonal
- Clone:9F10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Horse,Sheep,Non-human primates,Human,Bovine,Dog,Cat
- Form:Liquid
- Gene ID:UniprotID# P13612
- Antigen synonyms:CD49d|Antigen CD49D, alpha 4 subunit of VLA 4 receptor|CD49 antigen-like family member D|CD49 antigen like family member D|Integrin alpha IV|IA4|Integrin alpha-4|Integrin alpha 4 subunit|Integrin alpha 4
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, without Sodium Azide (0,2 µm filter sterilised). Endotoxin level is less than 0,01 EU/µg of the protein, as determined by the LAL test
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Unfortunately, this information is considered to be commercially sensitive
- Tested applications:IHC-Fr, Functional studies
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [9F10] antibody to Integrin alpha 4 for Flow Cytometry, IHC-Fr and Functional Studies with samples derived from Human, Bovine, Canine, Horse, Feline, Sheep and Non-Human Primates.
- Validated applications: Flow cytometry, IHC-Fr, Functional Studies
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: Integrin alpha 4
Clonality: Monoclonal
Clone: 9F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Bovine, Dog, Horse, Cat, Sheep, Non-Human Primates