Order Entry
Poland
ContactUsLinkComponent
Anti-Integrin alpha 4 Mouse Monoclonal Antibody [Clone: 9F10]
Anti-Integrin alpha 4 Mouse Monoclonal Antibody [Clone: 9F10]
Catalog # ANTIA86032-100
Supplier:  ANTIBODIES.COM
New Product
Anti-Integrin alpha 4 Mouse Monoclonal Antibody [Clone: 9F10]
Catalog # ANTIA86032-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    269C wild type
  • Antigen symbol:
    Integrin alpha 4
  • Clonality:
    Monoclonal
  • Clone:
    9F10
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Horse,
    Sheep,
    Non-human primates,
    Human,
    Bovine,
    Dog,
    Cat
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13612
  • Antigen synonyms:
    CD49d|Antigen CD49D, alpha 4 subunit of VLA 4 receptor|CD49 antigen-like family member D|CD49 antigen like family member D|Integrin alpha IV|IA4|Integrin alpha-4|Integrin alpha 4 subunit|Integrin alpha 4
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Unfortunately, this information is considered to be commercially sensitive
  • Tested applications:
    IHC-Fr
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [9F10] antibody to Integrin alpha 4 for Flow Cytometry and IHC-Fr with samples derived from Human, Bovine, Canine, Horse, Feline, Sheep and Non-Human Primates.

  • Validated applications: Flow cytometry, IHC-Fr
  • Recommended dilutions: IHC: 5 to 10 µg/ml

Type: Primary
Antigen: Integrin alpha 4
Clonality: Monoclonal
Clone: 9F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Bovine, Dog, Horse, Cat, Sheep, Non-Human Primates