Anti-CD18 Mouse Monoclonal Antibody [Clone: MEM-48] (PE (Phycoerythrin))
Catalog # ANTIA86016-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:95 subunit beta
- Antigen symbol:CD18
- Clonality:Monoclonal
- Clone:MEM-48
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P05107
- Antigen synonyms:CD18|Complement component 3 receptor 3 and 4 subunit|Complement receptor C3 beta subunit|CD 18|Beta 2 (antigen CD18 (p95)|Cell surface adhesion glycoprotein LFA 1/CR3/P150,959 beta subunit precursor)|Cell surface adhesion glycoproteins LFA-1/CR3/p150|Complement receptor C3 subunit beta|Cell surface adhesion glycoproteins LFA 1/CR3/p150,95 subunit beta
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Leukocytes from a patient suffering from a LGL Type leukemia
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [MEM-48] antibody to CD18 (PE) for Flow Cytometry with samples derived from Human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: MEM-48
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human