Anti-HLA DP + DQ + DR Mouse Monoclonal Antibody [clone: HLA-Pan/6663]
Catalog # ANTIA277868-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:HLA DP + DQ + DR
- Clonality:Monoclonal
- Clone:HLA-Pan/6663
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P04440, P01908, P01909, P01920, P01903
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:33 - 35 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Priess human B cell line.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [HLA-Pan/6663] antibody to HLA DP + DQ + DR for Flow Cytometry and IHC-P with samples derived from human.
- Validated Applications: Flow Cytometry, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HLA DP + DQ + DR
Clonality: Monoclonal
Clone: HLA-Pan/6663
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human