Anti-SPARC Mouse Monoclonal Antibody [clone: OSTN/3932]
Catalog # ANTIA277803-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:AA517111
- Antigen symbol:SPARC
- Clonality:Monoclonal
- Clone:OSTN/3932
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P09486
- Antigen synonyms:Cysteine rich protein|Basement membrane protein 40|Basement-membrane protein 40|BM 40|OI17|BM40|BM-40|MGC128090|hm:zeh0062
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:38 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 1-200, of human SPARC protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [OSTN/3932] antibody to SPARC for WB and IHC-P with samples derived from human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: OSTN/3932
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human