Anti-CD6 Rabbit Recombinant Antibody [clone: C6/2884R]
ANTIA253723-100
New Product
- Antibody type:Primary
- Antigen name:CD6
- Antigen symbol:CD6
- Clonality:Recombinant
- Clone:C6/2884R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P30203
- Antigen synonyms:CD6_HUMAN|CD6 antigen Tp120|T-cell differentiation antigen CD6|CD6 molecule|FLJ44171|T12|T cell differentiation antigen CD6|CD_antigen=CD6|OX52
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:90 - 130 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CD6 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [C6/2884R] antibody to CD6 for Flow Cytometry, IF and IHC-P with samples derived from human.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD6
Clonality: Recombinant
Clone: C6/2884R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human