Anti-CD5 Mouse Monoclonal Antibody [clone: C5/473 + CD5/54/F6]
ANTIA253711-100
New Product
- Antibody type:Primary
- Antigen name:CD 5
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:C5/473 + CD5/54/F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:Multiple
- Reactivity:Rabbit,Human,Bovine,Rat
- Form:Liquid
- Gene ID:UniprotID# P06127
- Antigen synonyms:CD5|CD5 molecule|CD5 antigen (p56 62)|CD5 antigen|LEU1|OTTHUMP00000236973|CD5_HUMAN|Lymphocyte antigen T1/Leu-1|LyA
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:67 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Clone C5/473: Recombinant human CD5 protein. Clone CD5/54/F6: A synthetic peptide from the intracellular region of human CD5 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [C5/473 + CD5/54/F6] antibody to CD5 for ELISA and IHC-P with samples derived from human, rabbit, bovine and rat.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: ELISA: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: C5/473 + CD5/54/F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human;Rabbit;Bovine;Rat