Anti-Collagen VI alpha 2 Rabbit Polyclonal Antibody
Catalog # ANTIA94735-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Typ przeciwciała:Primary
- Nazwa antygenu:CO6A2_HUMAN
- Symbol antygenu:Collagen VI alpha 2
- Klonalność:Poliklonalne
- Koniugacja:Unconjugated
- ELISA:Yes
- Gospodarz:Rabbit
- ImmunoChemia:Yes
- Izotyp:IgG
- Reaktywność:Human,Mouse
- Western blot:Yes
- Formularz:Liquid
- Identyfikator genu:UniprotID# P12110
- Synonimy antygenów:Collagen alpha 2(VI) chain|human mRNA for collagen VI alpha 2 C terminal globular domain|PP3610|Collagen alpha-2(VI) chain|collagen type VI alpha 2|Collagen VI alpha 2|COL6A2|Collagen VI alpha 2 polypeptide
- Bufor do przechowywania:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Waga molekularna:108 kDa
- Temperatura przechowywania:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Temperatura transportu:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human Collagen VI alpha2 (amino acids 691-740)
- Przetestowane aplikacje:IHC
- Oczyszczenie:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Rodzaj opakowania:Plastic vial
- Ilość:100 µG
Specifications
About this item
Rabbit polyclonal antibody to Collagen VI alpha2 for WB, IHC and ELISA with samples derived from Human and Mouse.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: Collagen VI alpha 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse