Order Entry
Poland
ContactUsLinkComponent
Anti-FOXN2 Rabbit Monoclonal Antibody [clone: ARC58450]
Anti-FOXN2 Rabbit Monoclonal Antibody [clone: ARC58450]
Catalog # ANTIA309852-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-FOXN2 Rabbit Monoclonal Antibody [clone: ARC58450]
Catalog # ANTIA309852-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    FOXN2
  • Antigen symbol:
    FOXN2
  • Clonality:
    Monoclonal
  • Clone:
    ARC58450
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P32314
  • Antigen synonyms:
    Forkhead box N2|Forkhead box protein N2|HTLF|Fkh19|Human T-cell leukemia virus enhancer factor|Foxn1|FOXN2|3230402J05Rik|FOXN2_HUMAN|6030465J18Rik
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% Proclin 300
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant protein of human FOXN2.
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC58450] antibody to FOXN2 for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: FOXN2
Clonality: Monoclonal
Clone: ARC58450
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human