Anti-CCL23 Mouse Monoclonal Antibody [clone: CCL23/4036]
ANTIA314570-100
New Product
- Antibody type:Primary
- Antigen name:C C motif chemokine ligand 23
- Antigen symbol:CCL23
- Clonality:Monoclonal
- Clone:CCL23/4036
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2c kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P55773
- Antigen synonyms:C-C motif chemokine 23|CCL23_HUMAN|CCL 23|Chemokine CC motif ligand 23|ck beta 8|CCL23(30-99)|Chemokine (C C motif) ligand 23|CCL23|C6 beta chemokine
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:13 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-120) of human CCL23 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [CCL23/4036] antibody to CCL23 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: CCL23
Clonality: Monoclonal
Clone: CCL23/4036
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human