Order Entry
Poland
ContactUsLinkComponent
Anti-Integrin alpha V Rabbit Monoclonal Antibody [clone: ARC50621]
Anti-Integrin alpha V Rabbit Monoclonal Antibody [clone: ARC50621]
Catalog # ANTIA308725-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Integrin alpha V Rabbit Monoclonal Antibody [clone: ARC50621]
Catalog # ANTIA308725-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    antigen identified by monoclonal
  • Antigen symbol:
    Integrin alpha V
  • Clonality:
    Monoclonal
  • Clone:
    ARC50621
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P06756
  • Antigen synonyms:
    Integrin alpha-5|CD 51|DKFZp686A08142|Integrin alpha-V light chain|CD51|integrin alpha V beta 3|Integrin alpha V|integrin alpha-V|Integrin alpha five
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% Proclin 300
  • Molecular weight:
    140 kDa
  • Sequence:
    SLKSSASFNVIEFPYKNLPIEDITNSTLVTTNVTWGIQPAPMPVPVWVIILAVLAGLLLLAVLVFVMYRMGFFKRVRPPQEEQEREQLQPHENGEGNSET
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 949-1048 of human Integrin alpha V (ITGAV/CD51) (NP_002201.2)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC50621] antibody to Integrin alpha V for WB, IHC and flow cytometry with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, flow cytometry
  • Recommended dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, flow cytometry: 1:50 to 1:200

Type: Primary
Antigen: Integrin alpha V
Clonality: Monoclonal
Clone: ARC50621
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat