Order Entry
Poland
ContactUsLinkComponent
Anti-CD45 Mouse Monoclonal Antibody [Clone: MEM-28] (PerCP (Peridinin-Chlorophyll Protein Complex))
Anti-CD45 Mouse Monoclonal Antibody [Clone: MEM-28] (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # ANTIA85886-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CD45 Mouse Monoclonal Antibody [Clone: MEM-28] (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # ANTIA85886-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B220
  • Antigen symbol:
    CD45
  • Clonality:
    Monoclonal
  • Clone:
    MEM-28
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08575
  • Antigen synonyms:
    CD45R|CD45|GP180|Leukocyte common antigen|L-CA|LCA|loc|CD45 antigen|CD 45
  • Storage buffer:
    Supplied in phosphate buffered saline with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human thymocytes and T lymphocytes
  • Purification:
    This antibody is conjugated with PerCP under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-28] antibody to CD45 (PerCP) for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: MEM-28
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human