Order Entry
Poland
ContactUsLinkComponent
 
Anti-CD34 Mouse Monoclonal Antibody [Clone: QBEnd-10] (PE (Phycoerythrin))
Catalog # ANTIA85663-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD34 Mouse Monoclonal Antibody [Clone: QBEnd-10] (PE (Phycoerythrin))
Catalog # ANTIA85663-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD34
  • Clonality:
    Monoclonal
  • Clone:
    QBEnd-10
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28906
  • Antigen synonyms:
    CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
  • Storage buffer:
    Supplied in phosphate buffered saline with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human endothelial vesicles
  • Purification:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [QBEnd-10] antibody to CD34 (PE) for flow cytometry with samples derived from human and non-human primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: QBEnd-10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates