Anti-HLA Class I Mouse Monoclonal Antibody [Clone: MEM-123]
Catalog # ANTIA85507-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:DKFZp686P19218
- Antigen symbol:HLA Class I
- Clonality:Monoclonal
- Clone:MEM-123
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG3
- Reactivity:Non-human primates,Human,Bovine
- Form:Liquid
- Antigen synonyms:EA2.1|HLA Class I|HLA CDA12|HLA 5.4|HLA A|HLA 6.2|HLA B|EA1.2|HLA C
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:COS-7 (African Green Monkey) cells
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MEM-123] antibody to HLA Class I for flow cytometry, IP and ELISA with samples derived from human, bovine and non-human primates.
- Validated applications: Flow cytometry, IP, ELISA
- Recommended dilutions: Flow cytometry: 4 µg/ml
Type: Primary
Antigen: HLA Class I
Clonality: Monoclonal
Clone: MEM-123
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Bovine, Non-Human Primates