Anti-CD8 Mouse Monoclonal Antibody [Clone: MEM-31] (PE (Phycoerythrin))
Catalog # ANTIA85482-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD8
- Clonality:Monoclonal
- Clone:MEM-31
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:OKT8 T cell antigen|CD8 antigen, alpha polypeptide (p32)|Leu2 T lymphocyte antigen|CD8A_HUMAN|CD8a|Leu2|MAL|CD8a antigen|CD8a molecule
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Crude thymus membrane fraction
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [MEM-31] antibody to CD8 (PE) for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: MEM-31
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human