Order Entry
Poland
ContactUsLinkComponent
Anti-CD8 Mouse Monoclonal Antibody [Clone: MEM-31] (APC (Allophycocyanin))
Anti-CD8 Mouse Monoclonal Antibody [Clone: MEM-31] (APC (Allophycocyanin))
Catalog # ANTIA85480-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD8 Mouse Monoclonal Antibody [Clone: MEM-31] (APC (Allophycocyanin))
Catalog # ANTIA85480-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8
  • Clonality:
    Monoclonal
  • Clone:
    MEM-31
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    Liquid
  • Antigen synonyms:
    OKT8 T cell antigen|CD8 antigen, alpha polypeptide (p32)|Leu2 T lymphocyte antigen|CD8A_HUMAN|CD8a|Leu2|MAL|CD8a antigen|CD8a molecule
  • Storage buffer:
    Supplied in phosphate buffered saline with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Crude thymus membrane fraction
  • Purification:
    This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-31] antibody to CD8 (APC) for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: MEM-31
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human