Anti-Htr1a Goat Polyclonal Antibody
ANTIA84977-100
New Product
- Antibody type:Primary
- Antigen name:5 HT 1A
- Antigen symbol:Htr1a
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:5 hydroxytryptamine (serotonin) receptor 1A|5-HT1A|5 hydroxytryptamine receptor 1A|5-HT-1A|5 HT1A|5-hydroxytryptamine receptor 1A|5 hydroxytryptamine (serotonin) receptor 1A G protein coupled|5 HT receptor 1A|5 HT1A receptor
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-ERKNERTAEAKRK
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Mouse Htr1a (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to HTR1A for ELISA and WB with samples derived from human and mouse.
- Validated applications: ELISA, W
Type: Primary
Antigen: Htr1a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse