Order Entry
Poland
ContactUsLinkComponent
Anti-KMT5A/SETD8/Pr-SET7 Rabbit Monoclonal Antibody [clone: ARC0916]
Anti-KMT5A/SETD8/Pr-SET7 Rabbit Monoclonal Antibody [clone: ARC0916]
Catalog # ANTIA307347-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-KMT5A/SETD8/Pr-SET7 Rabbit Monoclonal Antibody [clone: ARC0916]
Catalog # ANTIA307347-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    H4 K20 HMTase
  • Antigen symbol:
    KMT5A / SETD8 / Pr-SET7
  • Clonality:
    Monoclonal
  • Clone:
    ARC0916
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9NQR1
  • Antigen synonyms:
    Histone H4 K20 methyltransferase|N-lysine methyltransferase SETD8|KMT5A / SETD8 / Pr-SET7|Histone-lysine N-methyltransferase SETD8|Pr SET 7|H4-K20-HMTase SETD8|Lysine N-methyltransferase 5A|Histone lysine N methyltransferase H4 lysine 20 specific|H4 K20 specific histone methyltransferase
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    49 kDa
  • Sequence:
    MGEGGAAAALVAAAAAAAAAAAAVVAGQRRRRLGRRARCHGPGRAAGGKMSKPCAVEAAAAAVAATAPGPEMVERRGPGRPRTDGENVFTGQSKIYSYMS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 1-100 of human KMT5A/SETD8 (Q9NQR1).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC0916] antibody to KMT5A/SETD8/Pr-SET7 for WB with samples derived from Human and Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:1000

Type: Primary
Antigen: KMT5A / SETD8 / Pr-SET7
Clonality: Monoclonal
Clone: ARC0916
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse