Anti-Hsp90 alpha Rabbit Monoclonal Antibody [clone: ARC1167]
Catalog # ANTIA307595-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:EL52
- Antigen symbol:Hsp90 alpha
- Clonality:Monoclonal
- Clone:ARC1167
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P07900
- Antigen synonyms:Heat shock 86 kDa|heat shock 90kD protein, alpha-like 4|epididymis luminal secretory protein 52|heat shock 90kD protein 1, alpha|HS90A_HUMAN|Heat shock 90kDa protein 1 alpha|Heat shock protein HSP 90-alpha|Heat shock 90kD protein 1, alpha like 4|Heat shock protein 90kDa alpha (cytosolic) class A member 1
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:90 kDa
- Sequence:MPEETQTQDQPMEEEEVETFAFQAEIAQLMSLIINTFYSNKEIFLRELISNSSDALDKIRYESLTDPSKLDSGKELHINLIPNKQDRTLTIVDTGIGMTK
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Hsp90a (P07900)
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC1167] antibody to Hsp90 alpha for WB and IP with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IP
- Recommended Dilutions: WB: 1:500 to 1:2000, IP: 1:500 to 1:1000
Type: Primary
Antigen: Hsp90 alpha
Clonality: Monoclonal
Clone: ARC1167
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat