Order Entry
Poland
ContactUsLinkComponent
 
Anti-EGFR Rabbit Recombinant Antibody [clone: DM129]
New Product
 
undefined
Anti-EGFR Rabbit Recombinant Antibody [clone: DM129]
New Product

 

  • Catalog No:
  • ANTIA318578-500
  • ANTIA318578-100
  • Antigen symbol:
  • EGFR
  • EGFR
  • Antigen name:
  • 9030024J15Rik
  • 9030024J15Rik
  • Antigen synonyms:
  • EGFR_HUMAN|EGF R|EGFR|Avian erythroblastic leukemia viral (verbb) oncogene homolog|Cell growth inhibiting protein 40|AI552599|Epidermal growth factor receptor|Avian erythroblastic leukemia viral (v erb b) oncogene homolog|Cell proliferation inducing protein 61
  • EGFR_HUMAN|EGF R|EGFR|Avian erythroblastic leukemia viral (verbb) oncogene homolog|Cell growth inhibiting protein 40|AI552599|Epidermal growth factor receptor|Avian erythroblastic leukemia viral (v erb b) oncogene homolog|Cell proliferation inducing protein 61
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Recombinant
  • Recombinant
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • DM129
  • DM129
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • UniprotID# P00533
  • UniprotID# P00533
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Form:
  • Lyophilised
  • Lyophilised
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Concentration:
  • Reconstitution dependent
  • Reconstitution dependent
  • Storage buffer:
  • Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
  • Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
  • Storage temperature:
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C

 

Frequently Bought Together