Anti-Ctip1 / BCL-11A Rabbit Monoclonal Antibody [clone: ARC1486]
Catalog # ANTIA305976-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:Ctip1 / BCL-11A
- Clonality:Monoclonal
- Clone:ARC1486
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9H165
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:100 kDa
- Sequence:GEKPYKCNLCDHACTQASKLKRHMKTHMHKSSPMTVKSDDGLSTASSPEPGTSDLVGSASSALKSVVAKFKSENDPNLIPENGDEEEEEDDEEEEEEEEEEEEELTESERVDYGFGLSLEAARHHENSSRGAVVGVGDESRALPDVMQGMVLSSMQHFSEAFHQVLGEKHKRGHLAEAEGHRDTCDEDS
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 401-589 of human CTIP1/BCL11A (Q9H165).
- Tested applications:IHC
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC1486] antibody to Ctip1 / BCL-11A for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type: Primary
Antigen: Ctip1 / BCL-11A
Clonality: Monoclonal
Clone: ARC1486
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat