Order Entry
Poland
ContactUsLinkComponent
Anti-SPARC Rabbit Recombinant Antibody [clone: OSTN/8528R]
Anti-SPARC Rabbit Recombinant Antibody [clone: OSTN/8528R]
  ANTIA317057-100
New Product
Anti-SPARC Rabbit Recombinant Antibody [clone: OSTN/8528R]
  ANTIA317057-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    AA517111
  • Antigen symbol:
    SPARC
  • Clonality:
    Recombinant
  • Clone:
    OSTN/8528R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P09486
  • Antigen synonyms:
    Cysteine rich protein|Basement membrane protein 40|Basement-membrane protein 40|BM 40|OI17|BM40|BM-40|MGC128090|hm:zeh0062
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without Sodium azide and carrier free
  • Molecular weight:
    38 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 1-200) of human SPARC protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [OSTN/8528R] antibody to SPARC for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: SPARC
Clonality: Recombinant
Clone: OSTN/8528R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human