Order Entry
Poland
ContactUsLinkComponent
Anti-ADAM1B Rat Monoclonal Antibody [clone: #57]
Anti-ADAM1B Rat Monoclonal Antibody [clone: #57]
Catalog # ANTIA426-100
Supplier:  Antibodies.com
New Product
undefined
Anti-ADAM1B Rat Monoclonal Antibody [clone: #57]
Catalog # ANTIA426-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ADAM1B
  • Clonality:
    Monoclonal
  • Clone:
    #57
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8R534
  • Storage buffer:
    Supplied in phosphate buffered saline with 50% Glycerol (filter sterilised, without Sodium azide, and carrier free)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Mouse sperm
  • Tested applications:
    ICC, IHC-P
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rat monoclonal (#57) antibody to ADAM1B for ICC and IHC-P with samples derived from mouse.

  • Validated applications: ICC, IHC-P
  • Recommended dilutions: ICC: 1:100 to 1:300, IHC-P: 1:100 to 1:300

Type: Primary
Antigen: ADAM1B
Clonality: Monoclonal
Clone: #57
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse