Order Entry
Poland
ContactUsLinkComponent
Anti-IL-21 Recombinant Recombinant Antibody
Anti-IL-21 Recombinant Recombinant Antibody
Catalog # ANTIA318862-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-IL-21 Recombinant Recombinant Antibody
Catalog # ANTIA318862-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CVID11
  • Antigen symbol:
    IL-21
  • Clonality:
    Recombinant
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Recombinant
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Lyophilized
  • Gene ID:
    UniprotID# Q9HBE4
  • Antigen synonyms:
    IL21|Interleukin 21|Interleukin-21|IL-21|Interleukin 21 isoform|IL21_HUMAN|IL 21|Cytokine CX2|DN 452A22.7
  • Storage buffer:
    Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
  • Storage temperature:
    Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Concentration:
    Reconstitution dependent
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant humanized monoclonal [Avizakimab Biosimilar] antibody to IL-21 for ELISA with samples derived from human.

  • Validated applications: ELISA
  • Recommended dilutions: ELISA: 1:5000 to 10000

Type: Primary
Antigen: IL-21
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human