Anti-CD43 Mouse Monoclonal Antibody [clone: DF-T1]
Catalog # ANTIA253206-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 43
- Antigen symbol:CD43
- Clonality:Monoclonal
- Clone:DF-T1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P16150
- Antigen synonyms:Leukocyte large sialoglycoprotein|CD43 antigen|CD43|Galactoglycoprotein|Leucocyte sialoglycoprotein|GPL115|GPL 115|GALGP|Human gene for sialophorin
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:95/115/135 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Myeloblastic KG1 cells
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [DF-T1] antibody to CD43 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD43
Clonality: Monoclonal
Clone: DF-T1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human