Order Entry
Poland
ContactUsLinkComponent
Anti-beta 2 Microglobulin Mouse Monoclonal Antibody [clone: C21.48A1]
Anti-beta 2 Microglobulin Mouse Monoclonal Antibody [clone: C21.48A1]
Catalog # ANTIA252931-100
Supplier:  Antibodies.com
New Product
undefined
Anti-beta 2 Microglobulin Mouse Monoclonal Antibody [clone: C21.48A1]
Catalog # ANTIA252931-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B2M
  • Antigen symbol:
    beta 2 Microglobulin
  • Clonality:
    Monoclonal
  • Clone:
    C21.48A1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Non-human primates,
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P61769
  • Antigen synonyms:
    CDABP0092|Hdcma22p|Beta chain of mhc class 1 proteins|Beta chain of MHC class I molecules|Beta 2 microglobulin precursor|beta 2 Microglobulin|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin|B2MG_HUMAN
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    12 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Soluble beta 2 microglobulin
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [C21.48A1] antibody to beta 2 Microglobulin for Flow Cytometry, WB and IF with samples derived from Human and Non-Human Primates.

  • Validated applications: Flow cytometry, WB, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IF: 1 to 2 µg/ml

Type: Primary
Antigen: beta 2 Microglobulin
Clonality: Monoclonal
Clone: C21.48A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Non-Human Primates