Order Entry
Poland
ContactUsLinkComponent
Anti-Ki67 Rabbit Recombinant Antibody [clone: MKI67/4945R]
Anti-Ki67 Rabbit Recombinant Antibody [clone: MKI67/4945R]
Catalog # ANTIA252529-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Ki67 Rabbit Recombinant Antibody [clone: MKI67/4945R]
Catalog # ANTIA252529-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen identified by monoclonal
  • Antigen symbol:
    Ki67
  • Clonality:
    Recombinant
  • Clone:
    MKI67/4945R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P46013
  • Antigen synonyms:
    MIB 1|MIB|Antigen KI-67|MKI67|KIA|Marker of proliferation Ki-67|KI67_HUMAN|Antigen KI67|PPP1R105
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    345/395 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 2293 - 2478, of human Ki67 protein. The exact sequence is proprietary
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [MKI67/4945R] antibody to Ki67 for WB and IHC-P with samples derived from Human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Ki67
Clonality: Recombinant
Clone: MKI67/4945R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human