Order Entry
Poland
ContactUsLinkComponent
Anti-CD5 Mouse Monoclonal Antibody [clone: CD5/54/F6]
Anti-CD5 Mouse Monoclonal Antibody [clone: CD5/54/F6]
Catalog # ANTIA250530-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD5 Mouse Monoclonal Antibody [clone: CD5/54/F6]
Catalog # ANTIA250530-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD 5
  • Antigen symbol:
    CD5
  • Clonality:
    Monoclonal
  • Clone:
    CD5/54/F6
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Pig,
    Rabbit,
    Horse,
    Monkey,
    Chicken,
    Human,
    Bovine,
    Guinea pig,
    Rat
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P06127
  • Antigen synonyms:
    CD5|CD5 molecule|CD5 antigen (p56 62)|CD5 antigen|LEU1|OTTHUMP00000236973|CD5_HUMAN|Lymphocyte antigen T1/Leu-1|LyA
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    67 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide from the intracellular region of human CD5
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [CD5/54/F6] antibody to CD5 for IHC-P with samples derived from Human, Monkey, Horse, Porcine, Rabbit, Bovine, Rat, Guinea Pig and Chicken.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: CD5/54/F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Monkey;Horse;Porcine;Rabbit;Bovine;Rat;Guinea Pig;Chicken