Anti-CD45 Mouse Monoclonal Antibody [clone: 2B11 + PD7/26] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA251204-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:B220
- Antigen symbol:CD45
- Clonality:Monoclonal
- Clone:2B11 + PD7/26
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:Multiple
- Reactivity:Human,Dog
- Form:Liquid
- Gene ID:UniprotID# P08575
- Antigen synonyms:CD45R|CD45|GP180|Leukocyte common antigen|L-CA|LCA|loc|CD45 antigen|CD 45
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:180 - 220 kDa
- Storage temperature:Store at 4...8 °C. Antibody is stable for 24 months.
- Concentration:100 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Clone 2B11: Isolated neoplastic cells from T cell lymphoma. Clone PD7/26: Human peripheral blood lymphocytes maintained in T cell growth factor.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 T
Specifications
About this item
Mouse monoclonal [2B11 + PD7/26] antibody to CD45 (FITC) for Flow Cytometry, IF and IHC-P with samples derived from Human and Canine.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 2B11 + PD7/26
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human, Dog