Order Entry
Poland
ContactUsLinkComponent
Anti-CD44 Rabbit Recombinant Antibody [clone: HCAM/2875R]
Anti-CD44 Rabbit Recombinant Antibody [clone: HCAM/2875R]
Catalog # ANTIA250723-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD44 Rabbit Recombinant Antibody [clone: HCAM/2875R]
Catalog # ANTIA250723-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BA-1
  • Antigen symbol:
    CD44
  • Clonality:
    Recombinant
  • Clone:
    HCAM/2875R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P16070
  • Antigen synonyms:
    CD44_HUMAN|CD44|CDw44|CD44 molecule (Indian blood group)|chondroitin sulfate proteoglycan 8|CD 44|Cell surface glycoprotein CD44|CD44 antigen|CD44 molecule
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    80 - 95 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Stimulated human leukocytes
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [HCAM/2875R] antibody to CD44 for WB and IHC-P with samples derived from Human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 0,25 to 0,5 µg/ml

Type: Primary
Antigen: CD44
Clonality: Recombinant
Clone: HCAM/2875R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human