Order Entry
Poland
ContactUsLinkComponent
 
Anti-CD38 Mouse Recombinant Antibody [clone: rCD38/6447]
  ANTIA250695-100
New Product
undefined
Anti-CD38 Mouse Recombinant Antibody [clone: rCD38/6447]
  ANTIA250695-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Acute lymphoblastic leukemia cells antigen CD38
  • Antigen symbol:
    CD38
  • Clonality:
    Recombinant
  • Clone:
    rCD38/6447
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28907
  • Antigen synonyms:
    ADPRC 1|ADP-ribosyl cyclase 1|CD38 antigen|ADPRC1|cADPr hydrolase 1|ADP ribosyl cyclase/cyclic ADP-ribose hydrolase|ADP ribosyl cyclase|ADP ribosyl cyclase 1|CD 38
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    ~45 kDa (Glycoprotein)/35 kDa (protein core)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to amino acids within 200 - 300 of human CD38 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant mouse monoclonal [rCD38/6447] antibody to CD38 for WB and IHC-P with samples derived from Human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD38
Clonality: Recombinant
Clone: rCD38/6447
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human